C12H316CF643 Chemcia Scientific, LLC

2-(2-Chloro-pyridin-4-yl)-1,5,6,7-tetrahydro-pyrrolo[3,2-c]pyridin-4-one

  • MDL number MFCD12406067
  • Cas Number : 724726-05-0
  • Molecular Weight : 247.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2-chloro-4-pyridinyl)-1,5,6,7-tetrahydro-4H-pyrrolo[3,2-c]pyridin-4-one
Inchi Code 1S/C12H10ClN3O/c13-11-5-7(1-3-14-11)10-6-8-9(16-10)2-4-15-12(8)17/h1,3,5-6,16H,2,4H2,(H,15,17)
Inchi Key JTRXEPCDEVNYPE-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=14193

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14193
Availability details for
Quantity
  • Availability
  • Add to Cart