C12H316CF645 Chemcia Scientific, LLC

1-Bromo-5-chloro-4-fluoro-2-methoxy-benzene

  • MDL number MFCD18783171
  • Cas Number : 949892-08-4
  • Molecular Weight : 239.4727

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=14535

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14535
Availability details for
Quantity
  • Availability
  • Add to Cart