C12H316CF646 Chemcia Scientific, LLC

4-(Methanesulfonylamino-methyl)-benzoic acid

  • MDL number MFCD05855330
  • Cas Number : 696634-97-6
  • Molecular Weight : 229.25632

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=14536

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14536
Availability details for
Quantity
  • Availability
  • Add to Cart