C12H316CF64C Chemcia Scientific, LLC

5-Oxo-1,5-dihydro-pyrazolo[1,5-a]pyrimidine-3-carboxylic acid ethyl ester

  • Cas Number : 926663-00-5
  • Molecular Weight : 207.19038

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=14676

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14676
Availability details for
Quantity
  • Availability
  • Add to Cart