C12H316CF658 Chemcia Scientific, LLC

(4-Methoxy-3-nitro-phenyl)-carbamic acid tert-butyl ester

  • MDL number MFCD26841163
  • Cas Number : 1324000-28-3
  • Molecular Weight : 268.27172

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=14814

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14814
Availability details for
Quantity
  • Availability
  • Add to Cart