C12H316CF65A Chemcia Scientific, LLC

5-Bromo-2H-indazol-3-ylamine

  • Molecular Weight : 212.04997

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Gray Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=14822

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14822
Availability details for
Quantity
  • Availability
  • Add to Cart