C12H316CF65C Chemcia Scientific, LLC

8-Fluoro-3,4-dihydro-2H-benzo[1,4]oxazine hydrochloride

  • Molecular Weight : 189.61843

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=14826

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14826
Availability details for
Quantity
  • Availability
  • Add to Cart