C12H42134F7C Chemcia Scientific, LLC

2-Amino-5-iodo-4-methyl-nicotinic acid methyl ester

  • MDL number MFCD27918607
  • Molecular Weight : 292.07753

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5C
Shipping Temperature Ambient
Country of Origin USA
Product Link http://www.chemcia.com/product_fl02.asp?id=3007

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3007
Availability details for
Quantity
  • Availability
  • Add to Cart