C12H42134F80 Chemcia Scientific, LLC

6-Bromo-2H-indazol-3-ylamine

  • MDL number MFCD22543771
  • Cas Number : 404827-77-6
  • Molecular Weight : 212.04997

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4925

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4925
Availability details for
Quantity
  • Availability
  • Add to Cart