C12H42134F81 Chemcia Scientific, LLC

4-Bromo-2-chloro-benzothiazole-6-carboxylic acid methyl ester

  • MDL number MFCD28967994
  • Cas Number : 1403763-71-2
  • Molecular Weight : 306.5667

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5132

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5132
Availability details for
Quantity
  • Availability
  • Add to Cart