C12H42134F8A Chemcia Scientific, LLC

3-Amino-6-chloro-pyridin-2-ol

  • MDL number MFCD18257795
  • Cas Number : 1256808-08-8
  • Molecular Weight : 144.5614

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6708

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6708
Availability details for
Quantity
  • Availability
  • Add to Cart