C12H42134F8D Chemcia Scientific, LLC

6-Methyl-5-nitro-2-oxo-1,2-dihydro-pyridine-3-carbonitrile

  • MDL number MFCD00753863
  • Cas Number : 85216-54-2
  • Molecular Weight : 179.1362

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6985

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6985
Availability details for
Quantity
  • Availability
  • Add to Cart