C12H42134F90 Chemcia Scientific, LLC

Imidazo[1,2-a]pyrazine-2-carbaldehyde

  • MDL number MFCD09995736
  • Cas Number : 1017782-15-8
  • Molecular Weight : 147.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5C
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=7058

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7058
Availability details for
Quantity
  • Availability
  • Add to Cart