C12H42134F93 Chemcia Scientific, LLC

2-Amino-4-bromo-3-methyl-phenol

  • MDL number MFCD24646384
  • Cas Number : 1154740-46-1
  • Molecular Weight : 202.05191

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7829

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7829
Availability details for
Quantity
  • Availability
  • Add to Cart