C12H42134F94 Chemcia Scientific, LLC

1-Bromo-2-fluoro-4-iodo-5-methoxy-benzene

  • MDL number MFCD25976520
  • Cas Number : 1447671-70-6
  • Molecular Weight : 330.9241

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=8460

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=8460
Availability details for
Quantity
  • Availability
  • Add to Cart