C12H42134FA0 Chemcia Scientific, LLC

5-Methoxy-2H-pyrazolo[3,4-c]pyridine

  • Molecular Weight : 149.15334

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5C
Physical Form Light Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9170

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9170
Availability details for
Quantity
  • Availability
  • Add to Cart