C12H42134FA1 Chemcia Scientific, LLC

2-Bromo-7-chloro-5H-pyrrolo[2,3-b]pyrazine

  • MDL number MFCD12031300
  • Cas Number : 1150617-58-5
  • Molecular Weight : 232.46791

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9267

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9267
Availability details for
Quantity
  • Availability
  • Add to Cart