C12H42134FA2 Chemcia Scientific, LLC

3,5-Diiodo-6-methoxy-pyridin-2-ylamine

  • Molecular Weight : 375.93632

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9283

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9283
Availability details for
Quantity
  • Availability
  • Add to Cart