C12H42134FA3 Chemcia Scientific, LLC

1-(5-Methoxy-pyrazolo[3,4-c]pyridin-1-yl)-ethanone

  • Cas Number : 76006-02-5
  • Molecular Weight : 191.19098

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=9359

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=9359
Availability details for
Quantity
  • Availability
  • Add to Cart