C12H42134FA8 Chemcia Scientific, LLC

(2-Iodo-4-nitro-phenyl)-methanol

  • MDL number MFCD18397985
  • Cas Number : 500563-90-6
  • Molecular Weight : 279.03517

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10484

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10484
Availability details for
Quantity
  • Availability
  • Add to Cart