C12H42134FA9 Chemcia Scientific, LLC

(4-Amino-2-iodo-phenyl)-methanol

  • MDL number MFCD18389628
  • Cas Number : 1261645-45-7
  • Molecular Weight : 249.05231

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10485

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10485
Availability details for
Quantity
  • Availability
  • Add to Cart