C12H42134FAA Chemcia Scientific, LLC

6-Methoxy-3-methyl-benzo[b]thiophene-2-carboxylic acid

  • MDL number MFCD22490525
  • Cas Number : 23045-75-2
  • Molecular Weight : 222.26455

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=10540

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10540
Availability details for
Quantity
  • Availability
  • Add to Cart