C12H42134FB1 Chemcia Scientific, LLC

2,3-Dihydro-1H-pyrido[3,4-b][1,4]thiazine

  • MDL number MFCD28605149
  • Cas Number : 1547067-76-4
  • Molecular Weight : 152.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,3-dihydro-1H-pyrido[3,4-b][1,4]thiazine
Inchi Code 1S/C7H8N2S/c1-2-8-5-7-6(1)9-3-4-10-7/h1-2,5,9H,3-4H2
Inchi Key USYQMDLYUUZGQA-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=10858

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=10858
Availability details for
Quantity
  • Availability
  • Add to Cart