C12H42134FB4 Chemcia Scientific, LLC

2-Amino-3-iodo-5-trifluoromethyl-benzoic acid methyl ester

  • Cas Number : 1801782-87-5
  • Molecular Weight : 345.06124

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Pink Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11132

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11132
Availability details for
Quantity
  • Availability
  • Add to Cart