C12H42134FBF Chemcia Scientific, LLC

4-Methyl-6-oxo-1,6-dihydro-pyridine-3-carboxylic acid methyl ester

  • MDL number MFCD18836893
  • Cas Number : 1224465-02-4
  • Molecular Weight : 167.16583

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11368

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11368
Availability details for
Quantity
  • Availability
  • Add to Cart