C12H42134FC2 Chemcia Scientific, LLC

6-Fluoro-1H-[1,5]naphthyridin-4-one

  • Molecular Weight : 164.14025

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11430

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11430
Availability details for
Quantity
  • Availability
  • Add to Cart