C12H42134FC4 Chemcia Scientific, LLC

5-Methyl-3H-oxazolo[4,5-b]pyridin-2-one

  • MDL number MFCD13193501
  • Cas Number : 55656-25-2
  • Molecular Weight : 150.13807

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11650

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11650
Availability details for
Quantity
  • Availability
  • Add to Cart