C12H42134FC6 Chemcia Scientific, LLC

4-Bromo-2-methoxy-5-nitro-benzonitrile

  • Molecular Weight : 257.04465

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=11700

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=11700
Availability details for
Quantity
  • Availability
  • Add to Cart