C12H42134FCD Chemcia Scientific, LLC

2-Amino-3-chloro-5-iodo-benzoic acid methyl ester

  • Cas Number : 1070977-94-4
  • Molecular Weight : 311.50789

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=12746

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=12746
Availability details for
Quantity
  • Availability
  • Add to Cart