C12H42134FD7 Chemcia Scientific, LLC

4-Bromo-5-methoxy-2-nitro-benzaldehyde

  • MDL number MFCD23703341
  • Cas Number : 1196664-96-6
  • Molecular Weight : 260.04532

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=14054

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14054
Availability details for
Quantity
  • Availability
  • Add to Cart