C12H42134FD9 Chemcia Scientific, LLC

3-Iodo-5-trifluoromethyl-benzonitrile

  • MDL number MFCD12922961
  • Cas Number : 691877-05-1
  • Molecular Weight : 297.01941

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=14167

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14167
Availability details for
Quantity
  • Availability
  • Add to Cart