C12H42134FDB Chemcia Scientific, LLC

3-Bromo-5-fluoro-2H-indazole

  • MDL number MFCD28392831
  • Molecular Weight : 215.02573

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5C
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=14476

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14476
Availability details for
Quantity
  • Availability
  • Add to Cart