C12H42134FDD Chemcia Scientific, LLC

3-Chloro-6,7-dihydro-5H-pyrrolo[3,4-c]pyridazine maleate

  • Molecular Weight : 271.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-6,7-dihydro-5H-pyrrolo[3,4-c]pyridazine maleate
Inchi Code 1S/C6H6ClN3.C4H4O4/c7-6-1-4-2-8-3-5(4)9-10-6;5-3(6)1-2-4(7)8/h1,8H,2-3H2;1-2H,(H,5,6)(H,7,8)/b;2-1-
Inchi Key KSLVTJPYWAJAMP-BTJKTKAUSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=14830

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14830
Availability details for
Quantity
  • Availability
  • Add to Cart