C12H42134FDE Chemcia Scientific, LLC

2-Amino-3-bromo-5-trifluoromethyl-benzoic acid methyl ester

  • MDL number MFCD26960513
  • Cas Number : 1805624-33-2
  • Molecular Weight : 298.06084

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5C
Shipping Temperature Ambient
Country of Origin USA
Product Link http://www.chemcia.com/product_fl02.asp?id=14971

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14971
Availability details for
Quantity
  • Availability
  • Add to Cart