C12H42134FDF Chemcia Scientific, LLC

2-Amino-3-chloro-5-trifluoromethyl-benzoic acid methyl ester

  • MDL number MFCD28721371
  • Cas Number : 1806927-85-4
  • Molecular Weight : 253.60984

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=14972

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14972
Availability details for
Quantity
  • Availability
  • Add to Cart