C12H42134FE1 Chemcia Scientific, LLC

(S)-11-Formy-4-ethyl-4-hydroxy-1,12-dihydro-4H-2-oxa-6,12a-diaza-dibenzo[b,h]fluorene-3,13-dione

  • MDL number MFCD19441970
  • Cas Number : 80758-83-4
  • Molecular Weight : 376.37207

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=14968

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14968
Availability details for
Quantity
  • Availability
  • Add to Cart