C12H42134FE2 Chemcia Scientific, LLC

(S)-11-Dimethoxymethyl-4-ethyl-4-hydroxy-1,12-dihydro-4H-2-oxa-6,12a-diaza-dibenzo[b,h]fluorene-3,13-dione

  • MDL number MFCD19441969
  • Cas Number : 84017-99-2
  • Molecular Weight : 422.44159

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5C
Shipping Temperature Ambient
Country of Origin USA
Product Link http://www.chemcia.com/product_fl02.asp?id=14969

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14969
Availability details for
Quantity
  • Availability
  • Add to Cart