C12H42134FE5 Chemcia Scientific, LLC

1-Pyridin-2-yl-4,6-dihydro-1H-pyrrolo[3,4-c]pyrazole-5-carboxylic acid tert-butyl ester

  • Molecular Weight : 286.33631

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5C
Shipping Temperature Ambient
Country of Origin USA
Product Link http://www.chemcia.com/product_fl02.asp?id=14975

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14975
Availability details for
Quantity
  • Availability
  • Add to Cart