C12H42134FE6 Chemcia Scientific, LLC

2-Pyridin-2-yl-2,4,5,6-tetrahydro-pyrrolo[3,4-c]pyrazole trifluoro acetate

  • Molecular Weight : 300.24227

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=14976

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=14976
Availability details for
Quantity
  • Availability
  • Add to Cart