C12H42134FE7 Chemcia Scientific, LLC

2-Bromo-5-methyl-4-nitro-phenol

  • MDL number MFCD11101017
  • Cas Number : 14401-60-6
  • Molecular Weight : 232.03477

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=15156

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=15156
Availability details for
Quantity
  • Availability
  • Add to Cart