C12H42134FE8 Chemcia Scientific, LLC

3-Amino-6-fluoro-benzo[b]thiophene-2-carboxylic acid methyl ester

  • Molecular Weight : 225.24316

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=15200

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=15200
Availability details for
Quantity
  • Availability
  • Add to Cart