C12H580D8840 Chemcia Scientific, LLC

3-Bromo-6-nitro-[1,2,4]triazolo[4,3-a]pyridine

  • Molecular Weight : 243.02041

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=15619

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=15619
Availability details for
Quantity
  • Availability
  • Add to Cart