C12H580D8843 Chemcia Scientific, LLC

7-Bromo-2-chloromethyl-1H-quinazolin-4-one

  • MDL number MFCD27918625
  • Cas Number : 573681-17-1
  • Molecular Weight : 273.51797

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3145

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3145
Availability details for
Quantity
  • Availability
  • Add to Cart