C12H580D884A Chemcia Scientific, LLC

8-Chloro-[1,2,4]triazolo[1,5-a]pyrazin-2-ylamine

  • MDL number MFCD20727730
  • Cas Number : 1314874-80-0
  • Molecular Weight : 169.57413

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5017

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5017
Availability details for
Quantity
  • Availability
  • Add to Cart