C12H580D884B Chemcia Scientific, LLC

(S)-5-Chloro-N-((2-oxo-3-(4-(3-oxomorpholino)phenyl)-oxazolidin-5-yl)methyl )thiophene-2-carboxamide

  • MDL number MFCD11974010
  • Cas Number : 366789-02-8
  • Molecular Weight : 435.88941

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5191

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5191
Availability details for
Quantity
  • Availability
  • Add to Cart