C12H580D884F Chemcia Scientific, LLC

4-Bromo-5-fluoro-2-nitro-benzaldehyde

  • MDL number MFCD28739866
  • Cas Number : 213382-40-2
  • Molecular Weight : 248.00926

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5246

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5246
Availability details for
Quantity
  • Availability
  • Add to Cart