C12H580D8850 Chemcia Scientific, LLC

5-Iodo-4-trifluoromethyl-pyridin-2-ylamine

  • Cas Number : 1321530-57-7
  • Molecular Weight : 288.01178

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=5339

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=5339
Availability details for
Quantity
  • Availability
  • Add to Cart