C12H580D8857 Chemcia Scientific, LLC

5-tert-Butyl-2-chloro-phenylamine

  • MDL number MFCD11846051
  • Cas Number : 918445-56-4
  • Molecular Weight : 183.68278

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6133

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6133
Availability details for
Quantity
  • Availability
  • Add to Cart