C12H580D885B Chemcia Scientific, LLC

(5-Amino-2-nitro-phenyl)-methanol

  • MDL number MFCD00036068
  • Cas Number : 77376-03-5
  • Molecular Weight : 168.15341

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=6548

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=6548
Availability details for
Quantity
  • Availability
  • Add to Cart