C12H580D8860 Chemcia Scientific, LLC

(4-Fluoro-3-nitro-phenyl)-methyl-amine

  • MDL number MFCD17291755
  • Cas Number : 24773-12-4
  • Molecular Weight : 170.14444

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=7120

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=7120
Availability details for
Quantity
  • Availability
  • Add to Cart